Order Entry
Ireland
ContactUsLinkComponent
 
2,4,6-Trichloropyridine 98%
 
undefined
2,4,6-Trichloropyridine 98%

 

  • Appearance:
    White to light yellow Crystals or crystalline solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    31 to 37 °C
  • GC:
    ≥97.5 %

 

 

Frequently Bought Together