
3-(4-Bromophenyl)propionic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1643-30-7
:
Formula:C₉H₉BrO₂
MDL Number:MFCD01310793
:229.07 g/mol
Melting Pt:133…136 °C
Storage Temperature:Ambient
UN:3335
ADR:9,III
:GHS07
Specification Test Results
- Appearance:White to yellow to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:134 to 138 °C
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together