Order Entry
Ireland
ContactUsLinkComponent
Oxalic acid, anhydrous 98%
Oxalic acid, anhydrous 98%
Certificates
undefined
Oxalic acid, anhydrous 98%
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to almost white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    189 to 191 °C
  • Titration with NaOH:
    97.5 to 102.5 %
  • pH:
    8.5 to 10.5 (10 % suspension in water)

Specification Test Results

Related Information

Frequently Bought Together