


Tin(II) chloride, anhydrous 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7772-99-8
:
Formula:SnCl₂
MDL Number:MFCD00011241
:189.62 g/mol
Boiling Pt:623 °C (1013 hPa)
Melting Pt:246 °C
Density:3,95 g/cm³
UN:3260
ADR:8,III
:GHS07
:GHS08
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder or flakes
- Titration Iodimetric:97.5 to 102.5 %
- Iron (Fe):≤50 ppm
- Lead (Pb):≤75 ppm
Specification Test Results
Frequently Bought Together