Order Entry
Ireland
ContactUsLinkComponent
N,N-Diisopropylethylamine trihydrofluoride 90%
Toxic
Corrosive
Flammable
N,N-Diisopropylethylamine trihydrofluoride 90%
Certificates
undefined
Toxic
Corrosive
Flammable
N,N-Diisopropylethylamine trihydrofluoride 90%
Specification Test Results
  • Appearance:
    Clear colorless to yellow-brown Liquid
  • Infrared spectrum:
    Conforms
  • Titration with NaOH:
    18 to 22 % (available HF)