
3-Bromo-5-chloro-1,2,4-thiadiazole 97%
Catalog # ACRO297910100
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5965 to 1.5985 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
About this item
:
:199.45 g/mol
Boiling Pt:192…194 °C (760 mmHg)
Melting Pt:24…25 °C
:GHS07
Storage Temperature:Ambient
CAS Number:37159-60-7
Formula:C₂BrClN₂S
MDL Number:MFCD00799287