Order Entry
Ireland
ContactUsLinkComponent
Palladium, black 98%
Palladium, black 98%
Certificates
undefined
Palladium, black 98%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Black Powder
  • Assay:
    ≥97.5 % (Pd)
  • Calcium (Ca):
    ≤25 ppm
  • Copper (Cu):
    ≤25 ppm
  • Gold (Au):
    ≤50 ppm
  • Iridium (Ir):
    ≤50 ppm
  • Iron (Fe):
    ≤25 ppm
  • Lead (Pb):
    ≤25 ppm
  • Magnesium (Mg):
    ≤25 ppm
  • Platinum (Pt):
    ≤50 ppm
  • Rhodium (Rh):
    ≤50 ppm
  • Ruthenium (Ru):
    ≤50 ppm
  • Silicon (Si):
    ≤50 ppm
  • Silver (Ag):
    ≤50 ppm

Specification Test Results

Related Information

Frequently Bought Together