Order Entry
Ireland
ContactUsLinkComponent
2-(Aminomethyl)-1-ethylpyrrolidine 95%
Corrosive
Flammable
Irritant
2-(Aminomethyl)-1-ethylpyrrolidine 95%
Certificates
undefined
Corrosive
Flammable
Irritant
2-(Aminomethyl)-1-ethylpyrrolidine 95%

Specification Test Results

  • Appearance:
    Clear yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4655 to 1.4675 (20°C, 589 nm)
  • Titration with HClO4:
    ≥94 %
  • HPLC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together