Order Entry
Ireland
ContactUsLinkComponent
1,4-Dioxa-8-azaspiro[4.5]decane 98%
1,4-Dioxa-8-azaspiro[4.5]decane 98%
Certificates
undefined
1,4-Dioxa-8-azaspiro[4.5]decane 98%

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4809 to 1.4829 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Titration with HClO4:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together