
3-Butenoic acid 90% tech. unstabilised
:625-38-7
:
:C₄H₆O₂
:MFCD00002782
:86.09 g/mol
:163 °C (760 mmHg)
:–39 °C
:1,01 g/cm³
:65 °C
:Refrigerator
:3265
:8,III
:GHS05
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- GC:≥88 %
- Titration with NaOH:≥98.5 %
- Chloride (Cl):≤0.005 %
- Sulfate (SO4):≤0.005 %
- Heavy metals (as Pb):≤0.001 %
Frequently Bought Together