
Hexaketocyclohexane octahydrate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7255-28-9
:527-31-1
Formula:C₆O₆·8H₂O
MDL Number:MFCD00149074
:312.18 g/mol
Melting Pt:90…93 °C
Storage Temperature:Refrigerator
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Cream to brown Powder
- Infrared spectrum:Conforms
- Melting point:90 to 103 °C (Dec)
- Titration excess NaOH:≥96.0 %
Specification Test Results
Frequently Bought Together