Acid green 1, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:19381-50-1
:
Formula:C₃₀H₁₈FeN₃O₁₅S₃
MDL Number:MFCD00003886
:878.45 g/mol
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark green Crystalline powder
- Infrared spectrum:Conforms
- UV spectrum:Complies
Specification Test Results
Frequently Bought Together