Order Entry
Ireland
ContactUsLinkComponent
1-(2-Pyrimidyl)piperazine 98+%
Corrosive
1-(2-Pyrimidyl)piperazine 98+%
Certificates
undefined
Corrosive
1-(2-Pyrimidyl)piperazine 98+%

Specification Test Results

  • Appearance:
    Clear yellow Liquid after melting
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5850 to 1.5880 (20°C, 589 nm)
  • GC:
    ≥98.0 %
  • Residue after evaporation:
    ≤0.001 %

Specification Test Results

Related Information

Frequently Bought Together