Order Entry
Ireland
ContactUsLinkComponent
Trimethylphenylammonium hydroxide 0.1 M in methanol
Toxic
Health
Flammable
Trimethylphenylammonium hydroxide 0.1 M in methanol
Certificates
undefined
Toxic
Health
Flammable
Trimethylphenylammonium hydroxide 0.1 M in methanol

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Solution
  • Strength:
    0.09 to 0.11 M
  • Melting point:
    69 to 73 °C
  • GC:
    ≥98.0 %

Specification Test Results

Related Information

Frequently Bought Together