3-[2-(2-Aminoethylamino)ethylamino]propyltrimethoxysilane tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: N1-(2-Aminoethyl)-N2-(3-(trimethoxysilyl)propyl)ethane-1,2-diamine
CAS Number:35141-30-1
:
Formula:C₁₀H₂₇N₃O₃Si
MDL Number:MFCD00069173
Boiling Pt:114…118 °C (2 mmHg)
Density:1.031 g/cm³
Molecular Weight:265.429 g/mol
Flash Pt:137 °C
UN:3082
ADR:9 III
:GHS07
:GHS05
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid (May discolor to pink to red to brown on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4570 to 1.4650 (20°C, 589 nm)
- Titration with HClO4:14 to 18 % (as NH2)
Specification Test Results
Frequently Bought Together