(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (1R,2R)-(-)-1,2-Bis(methylamino)cyclohexane, (1R,2R)-N,N'-Dimethyl-1,2-cyclohexanediamine
CAS Number:68737-65-5
:
Formula:C₈H₁₈N₂
MDL Number:MFCD00671527
Molecular Weight:142.25 g/mol
Storage Temperature:Refrigerator
UN:1759
ADR:8 III
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to light yellow Low melting solid
- Infrared spectrum:Conforms
- GC:≥94 %
- Specific optical rotation:-137° to -149° (20°C, 589 nm) (C=1, CHCl3)
Specification Test Results
Frequently Bought Together