
(S)-(+)-γ-Hydroxymethyl-γ-butyrolactone 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:32780-06-6
:
Formula:C₅H₈O₃
MDL Number:MFCD00066224
Boiling Pt:110…115 °C (2 mmHg)
Density:1,23 g/cm³
Flash Pt:>110 °C
:116.12 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4680 to 1.4720 (20°C, 589 nm)
- GC:≥96.0 %
- Specific optical rotation:+30.5° to +36.5° (20°C, 589 nm) (c=3, EtOH)
Specification Test Results
Frequently Bought Together