
1-Adamantaneacetic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4942-47-6
:
Formula:C₁₂H₁₈O₂
MDL Number:MFCD00074728
:194.27 g/mol
Melting Pt:138…140 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:134 to 138 °C
- Titration with NaOH:≥97.5 %
Specification Test Results
Frequently Bought Together