
Verapamil hydrochloride 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:152-11-4
:23313-68-0
Formula:C₂₇H₃₈N₂O₄
MDL Number:MFCD00055208
:491.07 g/mol
Melting Pt:140…144 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:140 to 144 °C
- Loss on drying:≤0.5 % (105 °C, 2 h)
- Residue after ignition:≤0.1 %
- pH:4.5 to 6.5 (5 % solution)
- Titration with HClO4:99.0 to 100.5 % (on dried substance)
Specification Test Results
Frequently Bought Together