Methyl stearate mixture of homologs
:112-61-8
:
:CH₃(CH₂)₁₆CO₂CH₃
:MFCD00009005
:298.51 g/mol
:215 °C (15 mmHg)
:37…39 °C
:0,836 g/cm³
:>112 °C
:Ambient
:3,II
- Appearance:White to cream Waxy solid
- Infrared spectrum:Conforms
- Acid value:≤1 mg KOH/g
- Iodine value:≤1 g I2/100 g
- Saponification value:186 to 208 mg KOH/g
Frequently Bought Together