2-Ethylhexyl-4-methoxycinnamate 98% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5466-77-3
:
Formula:C₁₈H₂₆O₃
MDL Number:MFCD00072582
:290.40 g/mol
Density:1,009 g/cm³
Flash Pt:193 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5430 to 1.5470 (20°C, 589 nm)
- GC:≥97.5 %
- Stabilizer:0.05 to 0.1 % BHT
- Specific gravity:(25/25°C) 1.007 to 1.012
- Specific absorbance A (1%/1cm):≥830 (at 307 to 308 nm, MeOH)
- Acid value:≤1 mg KOH/g
Specification Test Results
Frequently Bought Together