Order Entry
Ireland
ContactUsLinkComponent
2-Amino-1-methylbenzimidazole 99+%
Irritant
2-Amino-1-methylbenzimidazole 99+%
Certificates
undefined
Irritant
2-Amino-1-methylbenzimidazole 99+%

Specification Test Results

  • Appearance:
    White to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    203 to 206 °C
  • GC:
    ≥99.0 %

Specification Test Results

Related Information

Frequently Bought Together