Anti-LFA3 Antibody
BSBTRP1106
- Antibody type:Primary
- Antigen name:CD58 molecule
- Antigen symbol:CD58
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:965
- Antigen synonyms:LFA-3|LFA3|ag3
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Lymphocyte function-associated antigen 3(CD58) detection. Tested with WB, IHC-P in Human.
Type: Primary
Antigen: CD58
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human