Order Entry
Ireland
ContactUsLinkComponent
Ficoll® PM400
Ficoll® PM400
Supplier:  Cytiva
Certificates
undefined
Ficoll® PM400
Supplier:  Cytiva
Certificates
Specifications
  • Catalog No:
  • 17-0300-50
  • 17-0300-10
  • 17-0300-05
  • Stability:
  • Stable in alkaline and neutral solutions, but are rapidly hydrolyzed in solution at pH 3, especially at elevated temperature
  • Stable in alkaline and neutral solutions, but are rapidly hydrolyzed in solution at pH 3, especially at elevated temperature
  • Stable in alkaline and neutral solutions, but are rapidly hydrolyzed in solution at pH 3, especially at elevated temperature