Order Entry
Ireland
ContactUsLinkComponent
Quantus®, Underwear T-Shirts
Quantus®, Underwear T-Shirts
Supplier:  VWR Collection
Certificates
undefined
Quantus®, Underwear T-Shirts
Supplier:  VWR Collection
Specifications
  • Catalog No:
  • 113-9184
  • 113-9183
  • 113-9182
  • 113-9181
  • QNTS151-5UWBU0XSID
  • 113-9185
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Risk area:
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Short
  • Short
  • Short
  • Short
  • Short
  • Short
  • Fabric:
  • Polyester Mikrofaser mit Karbon
  • Polyester Mikrofaser mit Karbon
  • Polyester Mikrofaser mit Karbon
  • Polyester Mikrofaser mit Karbon
  • Polyester Mikrofaser mit Karbon
  • Polyester Mikrofaser mit Karbon
  • Weight:
  • 97,5 g/m²
  • 97,5 g/m²
  • 97,5 g/m²
  • 97,5 g/m²
  • 97,5 g/m²
  • 97,5 g/m²