L-Leu-L-Ala, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Soluble in acetic acid 80% (50 mg/ml - clear colourless solution)
- Synonyms: Leu-Ala; H-Leu-Ala-OH
- Form: White Crystalline Powder
- Storage: 0 °C
- Optical Rotation: [α]20/D +19±3°, c = 2% in methanol
Used as fluorescence intensifier in fluorimetric methods for the determination of phenylalanine.
Specifications
- Protein/peptide name:L-Leu-L-Ala
Specifications
Frequently Bought Together