N-Acetyl-β-D-glucosamine
Catalog # SPCMA1269-25KGBL
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Fiber drum
- Pk:25 kg
- Seq:4283
Specifications
Specification Test Results
- Assay:99% Min.
Specification Test Results
About this item
:
:221.21 g/mol
Storage Temperature:Ambient
EINECS:231-368-2
CAS Number:7512-17-6
Formula:C₈H₁₅NO₆
MDL Number:MFCD00136044