Diisopropylamine
Supplier: Spectrum Chemical
Certificates
About this item
Synonyms: N-Isopropyl-2-propanamine
CAS Number:108-18-9
:
Formula:C₆H₁₅N
MDL Number:MFCD00008862
Boiling Pt:84
Melting Pt:–61 °C
Density:0.717 g/cm³ (20 °C)
Molecular Weight:101.19 g/mol
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:1158
ADR:3 II
Merck Index:13,03219
EINECS:203-558-5
:GHS02
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:99% Min.
Specification Test Results
Frequently Bought Together