Order Entry
Ireland
ContactUsLinkComponent
Vertical electrophoresis systems, omniPAGE mini wide
Vertical electrophoresis systems, omniPAGE mini wide
Supplier:  CLEAVER SCIENTIFIC
Certificates
undefined
Vertical electrophoresis systems, omniPAGE mini wide
Supplier:  CLEAVER SCIENTIFIC
Specifications
  • Catalog No:
  • CLEVVS10WDSYS
  • CLEVVS10WD
  • Model:
  • omniPAGE mini wide
  • omniPAGE mini wide
  • Gel size W×H:
  • 180×80 mm
  • 180×80 mm
  • Buffer volume, gel tank:
  • 600-2800 ml
  • 600-2800 ml
  • Glass plate size (W×H):
  • 200×100 mm
  • 200×100 mm
  • Unit W×H×D:
  • 260×160×160 mm
  • 260×160×160 mm