

2-Iodo-3-methylpyridine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:22282-58-2
:
Formula:C₆H₆IN
MDL Number:MFCD04039350
:219.03 g/mol
Boiling Pt:80 °C (0,3 mmHg)
:GHS06
:GHS07
Related Information
Frequently Bought Together