Order Entry
Ireland
ContactUsLinkComponent
4-[(1R)-2-Amino-1-hydroxyethyl]benzene-1,2-diol 98%
4-[(1R)-2-Amino-1-hydroxyethyl]benzene-1,2-diol 98%
Catalog # L08087.03
CAS Number:  
4-[(1R)-2-Amino-1-hydroxyethyl]benzene-1,2-diol 98%
Catalog # L08087.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:51-41-2
 :
Formula:C₈H₁₁NO₃
MDL Number:MFCD00025592
 :169.18 g/mol
Merck Index:14,6695
Melting Pt:∼ 220 °C (dec.)
EINECS:200-096-6
ADR:6.1,II
 :GHS06