1-Octen-3-one 97% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Oct-1-en-3-one, Pentyl Vinyl Ketone, Amyl Vinyl Ketone
CAS Number:4312-99-6
:
Formula:C₈H₁₄O
MDL Number:MFCD00036558
:126.20 g/mol
Boiling Pt:56…60 °C (16 mmHg)
Density:0,848
Flash Pt:35 °C (95 °F)
Storage Temperature:Refrigerator
EINECS:224-327-5
ADR:3,III
:Stabilised with BHA 0,1%
UN:1224
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together