Glycolaldehyde dimethyl acetal 98% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:30934-97-5
:
Formula:HOCH₂CH(OCH₃)₂
MDL Number:MFCD00051799
:106.12 g/mol
Boiling Pt:68 °C (21 mmHg)
Melting Pt:< –76 °C
Density:1,050
Flash Pt:66 °C (150 °F)
Storage Temperature:Ambient
EINECS:250-398-7
Related Information
Frequently Bought Together