Order Entry
Ireland
ContactUsLinkComponent
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) 99+% for molecular biology
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) 99+% for molecular biology
Catalog # J67233.22
CAS Number:  
undefined
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) 99+% for molecular biology
Catalog # J67233.22
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :157.6 g/mol
Melting Pt:∼ 152 °C (dec.)
Storage Temperature:Ambient
EINECS:214-684-5
CAS Number:1185-53-1
Formula:NH₂C(CH₂OH)₃·HCl
MDL Number:MFCD00012590