Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Gold
Gold 99% supplied in 0.1 mM PBS, nanoparticles 150 nm reactant-free
Gold 99% supplied in 0.1 mM PBS, nanoparticles 150 nm reactant-free
Gold 99% supplied in 0.1 mM PBS, nanoparticles 150 nm reactant-free
J67040.AC
:
Thermo Fisher Scientific
:
Gold 99% supplied in 0.1 mM PBS, nanoparticles 150 nm reactant-free
J67040.AC
:
Thermo Fisher Scientific
:
Pk:
25 mL
Seq:
2315
:
:
196.97 g/mol
:
Refrigerator
:
7440-57-5
:
Au
:
MFCD00003436