
Pefloxacine methanesulphonate dihydrate 95%
Catalog # J66285.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:333.4 · 96.1 · 36.0 g/mol
Merck Index:14,7066
Boiling Pt:529 °C
Melting Pt:271 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Refrigerator
EINECS:274-613-9
:GHS07
CAS Number:149676-40-4
Formula:C₁₇H₂₀FN₃O₃·CH₄O₃S·2H₂O
MDL Number:MFCD01685696