Prehybridisation solutions in SSC
Supplier: Thermo Fisher Scientific
Certificates
About this item
For the hybridization of probes to nucleic acids in Northern and Southern blotting.
Contains 6X SSC, 5X Denhardt's solution, 0,5% SDS, and 100 μg/ml of denatured DNA from salmon testes.
Applications: For the hybridization of probes to nucleic acids in Nothern and Southern blotting
Specifications
- Colour:Colourless
- Physical form:Liquid
Specifications
Related Information
Frequently Bought Together