Order Entry
Ireland
ContactUsLinkComponent
MOBS, 0,2 M buffer soln., pH 7,5
MOBS, 0,2 M buffer soln., pH 7,5
Catalog # J63519.AE
CAS Number:  
MOBS, 0,2 M buffer soln., pH 7,5
Catalog # J63519.AE
CAS Number:  

Specifications

  • Concentration:
    0,2 M
  • Pk:
    100 ml
  • CAS No.:
    115724-21-5
  • Description:
    MOBS buffer solution
  • MDL No.:
    MFCD00674916
  • Physical form:
    Liquid
  • pH value:
    7,5

Specifications

About this item

CAS No.: 115724-21-5
Physical Form: Liquid
MDL No.: MFCD00674916