Order Entry
Ireland
ContactUsLinkComponent
Zymogram renaturation buffer (10X)
Zymogram renaturation buffer (10X)
Catalog # J63137.AP
CAS Number:  
Zymogram renaturation buffer (10X)
Catalog # J63137.AP
CAS Number:  

Specifications

  • Pk:
    500 ml
  • Description:
    Zymogram renaturation buffer (10X)
  • Physical form:
    Liquid
  • Appearance:
    Colourless

Specifications

About this item

Physical Form: Liquid
Appearance: Colourless