
U-73122 95%
Catalog # J62898.MA
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:464.65 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:112648-68-7
Formula:C₂₉H₄₀N₂O₃
MDL Number:MFCD00893825