RIPA buffer with Triton® X-100 (5X)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Specifications
- Storage temperature:2°C to 8°C
- Physical form:Liquid
Specifications
Related Information
Frequently Bought Together