Order Entry
Ireland
ContactUsLinkComponent
 
KN-62
Catalog # J62444.MA
CAS Number:  
KN-62
Catalog # J62444.MA
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275
  • Environmentally Preferable:

Specifications

About this item

Synonyms: (S)-4-(2-(N-Methylisoquinoline-5-sulphonamido)-3-oxo-3-(4-phenylpiperazin-1-yl)propyl)phenyl isoquinoline-5-sulphonate
 :
 :721.84 g/mol
Storage Temperature:Freezer
CAS Number:127191-97-3
Formula:C₃₈H₃₅N₅O₆S₂
MDL Number:MFCD00083180
Melting Pt:65…67 °C