Agarose gel loading dye 6X for electrophoresis, alkaline
Agarose gel loading dye (6X), alkaline is used in DNA gel analysis in alkaline condition.
- 300 mm sodium hydroxide
- 6 mm EDTA
- 0,25% bromocresol green
- 0,25% xylene cyanol FF
- 18% Ficoll®
Used in agarose gel electrophoresis.
Frequently Bought Together