BLOTTO and Tween 20 in PBS
Catalog # J61880.AP
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:500 ml
- Description:BLOTTO and Tween 20 in PBS
- Storage temperature:-30°C to -10°C
- Physical form:Liquid
Specifications
About this item
Storage Temperature: −30 to −10 °C
Physical Form: Liquid
Applications: Blocking reagent in immunohistochemistry experiments