Order Entry
Ireland
ContactUsLinkComponent
Zymogram developing buffer (10X), pH 7,45
Zymogram developing buffer (10X), pH 7,45
Catalog # J61375.K2
CAS Number:  
Zymogram developing buffer (10X), pH 7,45
Catalog # J61375.K2
CAS Number:  

Specifications

  • Pk:
    1 L
  • Description:
    Zymogram developing buffer (10X)
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Viscous liquid
  • Odour/scent:
    Odourless
  • Appearance:
    Colourless
  • pH value:
    7.45

Specifications

About this item

Storage Temperature: 2 to 8 °C
Physical Form: Viscous liquid
Appearance: Colourless
Odour: Odourless