Order Entry
Ireland
ContactUsLinkComponent
 
Tricine-buffered saline (5X), pH 7,0
Catalog # J60811.AK
CAS Number:  
Tricine-buffered saline (5X), pH 7,0
Catalog # J60811.AK
CAS Number:  

Specifications

  • Concentration:
    5X
  • Pk:
    250 mL
  • CAS No.:
    5704-04-1
  • Description:
    Tricine-buffered saline
  • MDL No.:
    MFCD00004277
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless
  • Appearance:
    Colourless
  • pH value:
    7,0

Specifications

About this item

CAS No.: 5704-04-1
Physical Form: Liquid
Appearance: Colourless
Odour: Odourless
MDL No.: MFCD00004277