SSPE (20X), RNAse free
J60783.AP
:
- Pk:500 ml
- Description:SSPE (20X), RNAse free
- MDL No.:MFCD00132372
- Physical form:Liquid
SSPE (20X) is a standard reagent in Southern and Northern hybridisation procedures. This liquid can be used straight or diluted as needed.
Contains 0,2 M sodium phosphate, 3,0 M sodium chloride, 20 mM EDTA.