Order Entry
Ireland
ContactUsLinkComponent
 
HEPES-Buffered Saline, 5X Solutions
 
HEPES-Buffered Saline, 5X Solutions
 

 

  • CAS No.:
    7365-45-9
  • MDL No.:
    MFCD00006158
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless
  • Storage temperature:
    -30°C to -10°C
  • Appearance:
    Colourless

 

Frequently Bought Together