Order Entry
Ireland
ContactUsLinkComponent
TE buffer, (20X), pH 7,4, autoclaved
TE buffer, (20X), pH 7,4, autoclaved
Certificates
TE buffer, (20X), pH 7,4, autoclaved
Certificates

Specifications

  • MDL No.:
    MFCD00236359
  • Physical form:
    Liquid

Specifications

Related Information

Frequently Bought Together