Buffers, CAPS, 0.5 M solutions
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS No.: 1135-40-6
MDL No.: MFCD00003837
Specifications
- CAS No.:1135-40-6
- MDL No.:MFCD00003837
- Physical form:Liquid
Specifications
Related Information
Frequently Bought Together